1-Mercaptoadamantane, 98%
1-Mercaptoadamantane, 98%
1-Mercaptoadamantane, 98%
1-Mercaptoadamantane, 98%
Thermo Scientific Chemicals
1-Mercaptoadamantane, 98%
CAS: 34301-54-7 | C10H16S | 168.298 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ALFH58958.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material1-Mercaptoadamantane
CAS34301-54-7
Health Hazard 1H301-H319
Health Hazard 2GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c