4-(2-Pyridyl)benzaldehyde, 98%
4-(2-Pyridyl)benzaldehyde, 98%
4-(2-Pyridyl)benzaldehyde, 98%
4-(2-Pyridyl)benzaldehyde, 98%
Thermo Scientific Chemicals
4-(2-Pyridyl)benzaldehyde, 98%
CAS: 127406-56-8 | C12H9NO | 183.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFH55498.065 g
Catalog number ALFH55498.06
Price (MYR)
881.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
881.00
EA
Add to cart
Specifications
Chemical Name or Material4-(2-Pyridyl)benzaldehyde
CAS127406-56-8
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir
Solubility
Soluble in Chloroform
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Allison R. Dick.; Kami L. Hull.; Melanie S. Sanford. A Highly Selective Catalytic Method for the Oxidative Functionalization of C-H Bonds. J. Am. Chem. Soc. 2004, 126, (8), 2300-2301.
  2. Jun Han Lee.; Chang Su Lim.; Yu Shun Tian.; Ji Hee Han.; Bong Rae Cho. A Two-Photon Fluorescent Probe for Thiols in Live Cells and Tissues. J. Am. Chem. Soc. 2010, 132, (4), 1216-1217.