N-Boc-4-bromoaniline, 97%
N-Boc-4-bromoaniline, 97%
N-Boc-4-bromoaniline, 97%
N-Boc-4-bromoaniline, 97%
Thermo Scientific Chemicals
N-Boc-4-bromoaniline, 97%
CAS: 131818-17-2 | C11H14BrNO2 | 272.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFH55226.065 g
Catalog number ALFH55226.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Boc-4-bromoaniline
CAS131818-17-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c