N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals
N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals
N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals
N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH50256.031 g
Catalog number ALFH50256.03
Price (MYR)
2,066.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
2,066.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Isopropylpropylamine
CAS21968-17-2
Health Hazard 1H225-H314-H335
Health Hazard 2GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
N-Isopropylpropylamine is used as an active pharmaceutical ingredients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Isopropylpropylamine is used as an active pharmaceutical ingredients.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
N. C. Deno.; Richard E. FruitJr.. The oxidative cleavage of amines by aqueous bromine at 25.deg.. J. Am. Chem. Soc. 1968, 90, (13), 3502-3506.