Methyl blue
Methyl blue
Methyl blue
Thermo Scientific Chemicals
Methyl blue
CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol
Have Questions?
Catalog NumberQuantity
ALFH37721.1425 g
Catalog number ALFH37721.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl blue
CAS28983-56-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c