View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IRA-96 ion exchange resin
Physical FormBeads
CAS39409-19-3
Recommended StorageAmbient temperatures
TSCANo