Hydrogen chloride, 1M soln. in ethyl acetate
Hydrogen chloride, 1M soln. in ethyl acetate
Hydrogen chloride, 1M soln. in ethyl acetate
Hydrogen chloride, 1M soln. in ethyl acetate
Thermo Scientific Chemicals
Hydrogen chloride, 1M soln. in ethyl acetate
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
ALFH32266.AE100 mL
Catalog number ALFH32266.AE
Price (MYR)
733.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
733.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1H225-H315-H318-H335-H336
Health Hazard 2GHS H Statement
H225-H318-H332-H336-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if inhaled.
May cause drowsiness or dizziness.
Causes skin irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
View more
In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production.