3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH31635.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Chloro-4-(trifluoromethyl)benzaldehyde
CAS83279-38-3
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Chloro-4-(trifluoromethyl)benzaldehyde is used in synthesis of organic compunds in γ-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Philip J. Rybczynski,; Donald W. Combs,; Kimberly Jacobs,; Richard P. Shank,; Barry Dubinsky. γ-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines. J. Med. Chem. 1999, 42 (13),2403-2408.