D-(-)-Prolinamide, 98%
D-(-)-Prolinamide, 98%
D-(-)-Prolinamide, 98%
D-(-)-Prolinamide, 98%
Thermo Scientific Chemicals
D-(-)-Prolinamide, 98%
CAS: 62937-45-5 | C5H10N2O | 114.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFH30901.065 g
Catalog number ALFH30901.06
Price (MYR)
1,435.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,435.00
EA
Add to cart
Specifications
Chemical Name or MaterialD-(-)-Prolinamide
CAS62937-45-5
Melting Point100°C to 103°C
Recommended StorageAmbient temperatures
TSCANo
View more
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Zhuo Tang Dr.; Diana P. N. Gonçalves Dr.; Markus Wieland; Andreas Marx Prof. Dr.; and Jörg S. Hartig Prof. Dr. Novel DNA Catalysts Based on G-Quadruplex Recognition. ChemBioChem,2008, 9(7), 1061-1064.
  2. Gian Luigi Puleo; Anna Iuliano. Substrate control by means of the chiral cavity of prolinamide derivatives of cholic acid in the organocatalyzed Michael addition of cyclohexanone to nitroolefins. Tetrahedron: Asymmetry,2008, 19(17), 2045-2050.