N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
Thermo Scientific Chemicals
N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
CAS: 101555-62-8 | C20H19NO4 | 337.375 g/mol
Have Questions?
Catalog NumberQuantity
ALFH28500.065 g
Catalog number ALFH28500.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Fmoc-D-proline
CAS101555-62-8
Melting Point110°C to 116°C
Recommended StorageKeep cold
TSCANo
View more
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. asper Kaiser,; Sape S. Kinderman,; Bart C. J. van Esseveldt,; Floris L. van Delft,; Hans E. Schoemaker,; Richard H. Blaauwd,; Floris P. J. T. Rutjes. Synthetic Applications of Aliphatic Unsaturated α-H-α-Amino Acids.. Org. Biomol. Chem. 2005, 33435-3467 .
  2. Proline Derivatives and Analogs.Chemfiles. 512