1-Methyl-3-n-propylimidazolium iodide, 98%
1-Methyl-3-n-propylimidazolium iodide, 98%
1-Methyl-3-n-propylimidazolium iodide, 98%
1-Methyl-3-n-propylimidazolium iodide, 98%
Thermo Scientific Chemicals
1-Methyl-3-n-propylimidazolium iodide, 98%
CAS: 119171-18-5 | C7H13IN2 | 252.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27682.1850 g
Catalog number ALFH27682.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Methyl-3-n-propylimidazolium iodide
CAS119171-18-5
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362