1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
Thermo Scientific Chemicals
1-Butyl-2,3-dimethylimidazolium chloride, 99%
CAS: 98892-75-2 | C9H17ClN2 | 188.699 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27270.1850 g
Catalog number ALFH27270.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Butyl-2,3-dimethylimidazolium chloride
CAS98892-75-2
Melting Point89.5°C to 93.2°C
Recommended StorageAmbient temperatures
TSCANo