1-n-Hexyl-3-methylimidazolium chloride, 98%
1-n-Hexyl-3-methylimidazolium chloride, 98%
1-n-Hexyl-3-methylimidazolium chloride, 98%
1-n-Hexyl-3-methylimidazolium chloride, 98%
Thermo Scientific Chemicals
1-n-Hexyl-3-methylimidazolium chloride, 98%
CAS: 171058-17-6 | C10H19N2 | 167.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27178.1850 g
Catalog number ALFH27178.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-n-Hexyl-3-methylimidazolium chloride
CAS171058-17-6
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
RUO – Research Use Only
W Shi.; EJ Maginn. Atomistic Simulation of the Absorption of Carbon Dioxide and Water in the Ionic Liquid 1-n-Hexyl-3-methylimidazolium Bis (trifluoromethylsulfonyl) imide ([hmim][Tf2N]. The Journal of Physical Chemistry B, . 2008112 (7), 2045-2055.