1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%
1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%
1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%
1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%
Thermo Scientific Chemicals
1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%
CAS: 244193-52-0 | C12H23BF4N2 | 282.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27161.1850 g
Catalog number ALFH27161.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Methyl-3-n-octylimidazolium tetrafluoroborate
CAS244193-52-0
Recommended StorageAmbient temperatures
Density1.12
TSCANo