2-Chloro-L-phenylalanine, 98+%
2-Chloro-L-phenylalanine, 98+%
2-Chloro-L-phenylalanine, 98+%
2-Chloro-L-phenylalanine, 98+%
Thermo Scientific Chemicals
2-Chloro-L-phenylalanine, 98+%
CAS: 103616-89-3 | C9H10ClNO2 | 199.63 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27135.MD250 mg
Catalog number ALFH27135.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material2-Chloro-L-phenylalanine
Melting Point196°C to 199°C
CAS103616-89-3
Recommended StorageAmbient temperatures
TSCANo
View more
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Robert Opitza,; Matthias Müllera,; Cédric Reuterb,; Matthias Baronea,; Arne Soickeb,; Yvette Roskec,; Kirill Piotukh. A modular toolkit to inhibit proline-rich motif-mediated protein-protein interactions. Journal name. 2015, 112(16) ,5011-5016.