Ammonia, 2M in methanol
Ammonia, 2M in methanol
Ammonia, 2M in methanol
Ammonia, 2M in methanol
Thermo Scientific Chemicals
Ammonia, 2M in methanol
CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27080.AE100 mL
Catalog number ALFH27080.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonia
CAS7664-41-7
Health Hazard 1H225-H301+H311+H331-H315-H318-H336-H370-H372
Health Hazard 2GHS H Statement
H225-H311-H331-H370-H318-H315
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes serious eye damage.
Causes skin irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
View more
Ammonia, 2M in methanol is used as a solvent in organic reactions and as a mobile phase in thin layer chromatography.