(+/-)-Propranolol hydrochloride, 99%
(+/-)-Propranolol hydrochloride, 99%
(+/-)-Propranolol hydrochloride, 99%
(+/-)-Propranolol hydrochloride, 99%
Thermo Scientific Chemicals
(+/-)-Propranolol hydrochloride, 99%
CAS: 318-98-9 | C16H22ClNO2 | 295.807 g/mol
Have Questions?
Catalog NumberQuantity
ALFH26645.065 g
Catalog number ALFH26645.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-Propranolol hydrochloride
CAS318-98-9
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
A beta-adrenergic blocker