4-n-Hexylaniline, 98%
4-n-Hexylaniline, 98%
4-n-Hexylaniline, 98%
4-n-Hexylaniline, 98%
Thermo Scientific Chemicals
4-n-Hexylaniline, 98%
CAS: 33228-45-4 | C12H19N | 177.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH26297.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-n-Hexylaniline
CAS33228-45-4
Health Hazard 1H302-H315-H319
Health Hazard 2GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
View more
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
RUO – Research Use Only
General References:
  1. Zhengjin Jiang et. al. Mixed-mode reversed-phase and ion-exchange monolithic columns for micro-HPLC.. Journal of Separation Science. 2008 , 31 (15) ,2774-2783.
  2. Lioubov I Brueggemann.; Jack M Sullivan.HEK293S cells have functional retinoid processing machinery.. Journal of General Physiology. 2002, 119 (6), 593-612.