Phloroglucinol, anhydrous, 98%
Phloroglucinol, anhydrous, 98%
Phloroglucinol, anhydrous, 98%
Phloroglucinol, anhydrous, 98%
Thermo Scientific Chemicals
Phloroglucinol, anhydrous, 98%
CAS: 108-73-6 | C6H6O3 | 126.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFB25502.30
Price (MYR)
1,594.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
1,594.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhloroglucinol
Name Noteanhydrous
CAS108-73-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .
Solubility
Soluble in diethyl ether, ethanol and pyridine.
Notes
Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
RUO – Research Use Only
General References:
  1. Reacts with mono- or dimethylamine in DMF-water, with displacement of one of the OH groups, to give 5-aminoresorcinols: Helv. Chim. Acta, 56, 510 (1973).
  2. The electron-rich nucleus undergoes acylation with nitriles under mild Lewis acid catalysis, to give ketones (Houben-Hoesch reaction).
  3. Selective monobromination cannot be achieved conventionally. For bromination-debromination protocols for the preparation of the mono- and di-brominated derivatives, see: Can. J. Chem., 67, 335 (1989).
  4. Dubey, R.; Desiraju, G. R. Combinatorial Crystal Synthesis: Structural Landscape of Phloroglucinol: 1, 2-bis (4-pyridyl) ethylene and Phloroglucinol: Phenazine. Angew. Chem. Int. Ed. 2014, 53 (48), 13178-13182.
  5. Ghimbeu, C. M.; Vidal, L.; Delmotte, L.; Le Meins, J. M.; Vix-Guterl, C. Catalyst-free soft-template synthesis of ordered mesoporous carbon tailored using phloroglucinol/glyoxylic acid environmentally friendly precursors. Green Chem. 2014, 16 (6), 3079-3088.