Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™
Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™
Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™
Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™
Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFB24998.065 g
Catalog number ALFB24998.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropyl N-phenylcarbamate
CAS122-42-9
Health Hazard 1H302
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.