4-Fluorosalicylic acid, 98%
4-Fluorosalicylic acid, 98%
4-Fluorosalicylic acid, 98%
4-Fluorosalicylic acid, 98%
Thermo Scientific Chemicals
4-Fluorosalicylic acid, 98%
CAS: 345-29-9 | C7H5FO3 | 156.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFB24992.06
Price (MYR)
849.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
849.00
EA
Add to cart
Specifications
Chemical Name or Material4-Fluorosalicylic acid
CAS345-29-9
Health Hazard 1H302-H315-H319-H335-H500
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Terézia Szabó-Plánka; Ján Moncol; Eszter Tóth; Béla Gyurcsik; Nóra Veronika Nagy; Zuzana Vasková; Antal Rockenbauer; Dušan Valigura. ESR and pH-potentiometric study of the mixed-ligand complex formation in the copper(II)-4-fluorosalicylic acid-N,N-diethylnicotinamide system: Structure and spectral properties of [Cu(4-fluorosalicylate)2(N,N-diethylnicotinamide)2(H2O)2] complex. Polyhedron.2011, 30, 2421-2429.