5-Nitroisatin, 98%
5-Nitroisatin, 98%
5-Nitroisatin, 98%
Thermo Scientific Chemicals
5-Nitroisatin, 98%
CAS: 611-09-6 | C8H4N2O4 | 192.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24962.065 g
Catalog number ALFB24962.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Nitroisatin
CAS611-09-6
Health Hazard 1H302-H312-H332-H340
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c