Isoeugenol, cis + trans, 98%
Isoeugenol, cis + trans, 98%
Isoeugenol, cis + trans, 98%
Isoeugenol, cis + trans, 98%
Thermo Scientific Chemicals
Isoeugenol, cis + trans, 98%
CAS: 97-54-1 | C10H12O2 | 164.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24541.22100 g
Catalog number ALFB24541.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIsoeugenol
Name Notecis + trans
CAS97-54-1
Health Hazard 1H302-H315-H317-H319
Health Hazard 2GHS H Statement
H302-H315-H319-H335