2-Aminoterephthalic acid, 99%
2-Aminoterephthalic acid, 99%
2-Aminoterephthalic acid, 99%
2-Aminoterephthalic acid, 99%
Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%
CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB24441.14
Price (MYR)
577.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
577.00
EA
Add to cart
Specifications
Chemical Name or Material2-Aminoterephthalic acid
CAS10312-55-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Polya M Miladinova and Temenushka N Konstantinova. Synthesis and properties of some new blue-emitting triazine derivatives of 2-aminoterephthalic acid, containing a polymerisable group and stabiliser fragment. Coloration Technology. 2009, 125 (4), 242-247.
  2. J. Arias-Pardilla; H.J. Salavagione; C. Barbero; E. Morallón; J.L. Vázquez. Study of the chemical copolymerization of 2-aminoterephthalic acid and aniline.: Synthesis and copolymer properties. European Polymer Journal. 2006, 42 (7), 1521-1532.