3,4-Diaminotoluene, 97%
3,4-Diaminotoluene, 97%
3,4-Diaminotoluene, 97%
3,4-Diaminotoluene, 97%
Thermo Scientific Chemicals
3,4-Diaminotoluene, 97%
CAS: 496-72-0 | C7H10N2 | 122.171 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24378.22100 g
Catalog number ALFB24378.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3,4-Diaminotoluene
CAS496-72-0
Health Hazard 1H302-H319-H341
Health Hazard 2GHS H Statement
H301-H341-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye irritation.
Health Hazard 3P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c