N-Acetyl-L-methionine, 99%
N-Acetyl-L-methionine, 99%
N-Acetyl-L-methionine, 99%
N-Acetyl-L-methionine, 99%
Thermo Scientific Chemicals
N-Acetyl-L-methionine, 99%
CAS: 65-82-7 | C7H13NO3S | 191.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24312.065 g
Catalog number ALFB24312.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Acetyl-L-methionine
Melting Point103°C to 107°C
CAS65-82-7
Recommended StorageAmbient temperatures
RTECS NumberPD0480000