Benzenesulfonyl hydrazide, 98%
Benzenesulfonyl hydrazide, 98%
Benzenesulfonyl hydrazide, 98%
Benzenesulfonyl hydrazide, 98%
Thermo Scientific Chemicals
Benzenesulfonyl hydrazide, 98%
CAS: 80-17-1 | C6H8N2O2S | 172.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23971.1425 g
Catalog number ALFB23971.14
Price (MYR)
350.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
350.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzenesulfonyl hydrazide
CAS80-17-1
Health Hazard 1H228-H242
Health Hazard 2GHS H Statement
H242-H228-H301
Heating may cause a fire.
Flammable solid.
Toxic if swallowed.
Health Hazard 3P210-P220-P234-P240-P241-P280-P370+P378q-P375-P380-P501c