Heptanal, 97%
Heptanal, 97%
Heptanal, 97%
Thermo Scientific Chemicals
Heptanal, 97%
CAS: 111-71-7 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number ALFB23830.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHeptanal
CAS111-71-7
Health Hazard 1H226-H315
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
View more
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
Solubility
Soluble in water 1.3 g/L.
Notes
Hygroscopic. Store away from oxidizing agents, air.
RUO – Research Use Only
General References:
  1. Didier Tichit; Doina Lutic; 1, Bernard Coq; Robert Durand; Rémy Teissier. The aldol condensation of acetaldehyde and heptanal on hydrotalcite-type catalysts. Journal of Catalysis. 2003, 219 (1), 167-175.
  2. Suzanne E. Paulson; De-Ling Liu and Grazyna E. Photolysis of Heptanal. J. Org. Chem. 2006, 71 (17), 6403-6408.