2-Hydroxy-3-nitrobenzaldehyde, 98%
2-Hydroxy-3-nitrobenzaldehyde, 98%
2-Hydroxy-3-nitrobenzaldehyde, 98%
2-Hydroxy-3-nitrobenzaldehyde, 98%
Thermo Scientific Chemicals
2-Hydroxy-3-nitrobenzaldehyde, 98%
CAS: 5274-70-4 | C7H5NO4 | 167.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23658.065 g
Catalog number ALFB23658.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Hydroxy-3-nitrobenzaldehyde
CAS5274-70-4
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c