Tin(II) 2-ethylhexanoate, 95%
Tin(II) 2-ethylhexanoate, 95%
Tin(II) 2-ethylhexanoate, 95%
Tin(II) 2-ethylhexanoate, 95%
Thermo Scientific Chemicals
Tin(II) 2-ethylhexanoate, 95%
CAS: 301-10-0 | C16H30O4Sn | 405.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB23612.22
Price (MYR)
602.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
602.00
EA
Add to cart
Specifications
Chemical Name or MaterialTin(II) 2-ethylhexanoate
CAS301-10-0
Health Hazard 1H317-H318-H361
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
View more
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Olsen, T. R.; Davis, L. L.; Nicolau, S. E.; Duncan, C. C.; Whitehead, D. C.; Van Horn, B. A.; Alexis, F. Non-invasive deep tissue imaging of iodine modified poly (caprolactone-co-1-4-oxepan-1, 5-dione) using X-ray. Acta Biomater. 2015, 20, 94-103.
  2. Sosnowski, S.; Lewinski, P. L-Lactide polymerization catalysed by tin(II) 2-ethyl-hexanoate. A deeper look at chain transfer reactions. Polym. Chem. 2015, 6 (35), 6292-6296.