2-Chloro-3',4'-dihydroxyacetophenone, 97%
2-Chloro-3',4'-dihydroxyacetophenone, 97%
2-Chloro-3',4'-dihydroxyacetophenone, 97%
2-Chloro-3',4'-dihydroxyacetophenone, 97%
Thermo Scientific Chemicals
2-Chloro-3',4'-dihydroxyacetophenone, 97%
CAS: 99-40-1 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23496.1425 g
Catalog number ALFB23496.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Chloro-3',4'-dihydroxyacetophenone
CAS99-40-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
Solubility
Soluble in DMSO, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
Huidobro-Toro JP; Carpi A. Selective blocking properties of chloro acetyl catechol (CAC) on the alpha-adrenergic receptor. Europe PMC. 1976, 222 (2), 180-192.