2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
Thermo Scientific Chemicals
2-Ethylhexanal, 96%
CAS: 123-05-7 | C8H16O | 128.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFB23471.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethylhexanal
CAS123-05-7
Health Hazard 1H226-H317-H361
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P210-P233-P240-P241-P242-P243-P261-P272-P280g-P281-P303+P361+P353-P308+P313-P333+P313-P363-P370+P378q-P501c
View more
2-Ethylhexanal is used as an intermediate for chemical synthesis and in the production of pharmaceuticals and odorous substances.  
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexanal is used as an intermediate for chemical synthesis & in the production of disinfectants, pharmaceuticals and odorous substances.  
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Christel Lehtinen and Gösta Brunow. Epoxidation of Terminal Alkenes with Oxygen and 2-Ethyl Hexanal, without Added Catalyst or Solvent. Org. Proc. Res. Dev. 1999, 3, (2), 101-103.
  2. Christel Lehtinen and Gösta Brunow. Factors Affecting the Selectivity of Air Oxidation Of 2-Ethyhexanal, an α-Branched Aliphatic Aldehyde. Org. Proc. Res. Dev. 2000, 4 (6), 544-549.