D-Norvaline, 99%
D-Norvaline, 99%
D-Norvaline, 99%
Thermo Scientific Chemicals
D-Norvaline, 99%
CAS: 2013-12-9 | C5H11NO2 | 117.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23444.031 g
Catalog number ALFB23444.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Norvaline
Melting Point>300°C
CAS2013-12-9
Recommended StorageAmbient temperatures
EINECS Number217-936-2