1-Chlorohexadecane, 97%
1-Chlorohexadecane, 97%
Thermo Scientific Chemicals
1-Chlorohexadecane, 97%
CAS: 4860-03-1 | C9H7F3O3 | 220.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23406.AE100 mL
Catalog number ALFB23406.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Chlorohexadecane
CAS4860-03-1
Melting Point8°C to 10°C
Recommended StorageAmbient temperatures
Density0.865
View more
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.
Solubility
Soluble in ether. Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. G L Murphy and J J Perry. Chlorinated fatty acid distribution in Mycobacterium convolutum phospholipids after growth on 1-chlorohexadecane. Appl. Environ. Microbiol. 1987, 53 (1), 10-13.
  2. Masamichi Akimoto; Hideharu Tanaka and Akiko Watanabe. Dechlorination of 1-chlorohexadecane and 2-chloronaphthalene in water under sub- and supercritical conditions. The Canadian Journal of Chemical Engineering. 2000, 78 (6), 1151-1156.