Cobalt(II) acetate, anhydrous, 98+%
Cobalt(II) acetate, anhydrous, 98+%
Cobalt(II) acetate, anhydrous, 98+%
Cobalt(II) acetate, anhydrous, 98+%
Thermo Scientific Chemicals
Cobalt(II) acetate, anhydrous, 98+%
CAS: 71-48-7 | C4H6CoO4 | 177.02 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23218.1850 g
Catalog number ALFB23218.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II) acetate
Name Noteanhydrous
CAS71-48-7
Health Hazard 1H317-H334-H341-H350i-H360F
Health Hazard 2GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
View more
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.
Solubility
Soluble in water, alcohol and dilute acids.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Rajabi, F.; Feiz, A.; Luque, R. An Efficient Synthesis of Coumarin Derivatives Using a SBA-15 Supported Cobalt(II) Nanocatalyst. Catal. Lett. 2015, 145 (8), 1621-1625.
  2. Xu, D.; Xie, Y.; Song, Y. J.; Deng, W. Q. A green and facile method toward synthesis of waste paper-derived 3D functional porous graphene via in situ activation of cobalt(II). J. Mater. Chem. A 2015, 3 (31), 16072-16078.