3-Dimethylaminophenol, 97+%
3-Dimethylaminophenol, 97+%
3-Dimethylaminophenol, 97+%
3-Dimethylaminophenol, 97+%
Thermo Scientific Chemicals
3-Dimethylaminophenol, 97+%
CAS: 99-07-0 | C8H11NO | 137.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23067.22100 g
Catalog number ALFB23067.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Dimethylaminophenol
CAS99-07-0
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents, light.
RUO – Research Use Only
General References:
  1. Bozena Bukowska; Jaromir Michałowicz; Wirgiliusz Duda. Alterations in human red blood cell properties induced by 3-(dimethylamino)phenol (in vitro). Toxicology in Vitro. 2007, 21, (8), 1574-1580.
  2. Kling RE. UV spectrophotometric assay of neostigmine methylsulfate as the alkaline hydrolysis product, 3-dimethylaminophenol. J Assoc Off Anal Chem. 1971, 54 (1), 21-6.