Zinc gluconate hydrate, 97%
Zinc gluconate hydrate, 97%
Zinc gluconate hydrate, 97%
Zinc gluconate hydrate, 97%
Thermo Scientific Chemicals
Zinc gluconate hydrate, 97%
CAS: 4468-02-4 | C4H7NO | 85.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23022.22100 g
Catalog number ALFB23022.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc gluconate hydrate
CAS4468-02-4
Recommended StorageAmbient temperatures
RTECS NumberZH3750000
pH6.5
View more
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.
Solubility
Soluble in water 100g/ l.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Sunil Sazawal; Margaret Bentley; Robert E. Black; Pratibha Dhingra; Sherly George; Maharaj K. Bhan. Effect of Zinc Supplementation on Observed Activity in Low Socioeconomic Indian Preschool Children. Pediatrics. 1996, 98 (1), 1132 -1137.
  2. L Saghaie; GH Houshfar; M Neishabor. Synthesis and Determination of Partition Coefficients of Zinc Complexes with Clinical Potential application. IJPR. 2006, 5 (3), 179-189.