Dimethyl suberate, 99%
Dimethyl suberate, 99%
Dimethyl suberate, 99%
Dimethyl suberate, 99%
Thermo Scientific Chemicals
Dimethyl suberate, 99%
CAS: 1732-09-8 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB22681.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl suberate
CAS1732-09-8
Recommended StorageAmbient temperatures
Density1.016
EINECS Number257-059-5
View more
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. D.H.Nugteren. Arachidonate lipoxygenase in blood platelets. Biochimica et Biophysica Acta (BBA) - Lipids and Lipid Metabolism. 1975, 380,(2), 299-307.
  2. A.P.Tulloch. Synthesis of [11-2H2], [8-2H2], [7-2H2], [6-2H2], [5-2H2], [4-2H2] and [3-2H2] cis-9-octadecenoates. Chemistry and Physics of Lipids. 1979, 25,(3), 225-235.