Polyethylene glycol 600
Polyethylene glycol 600
Polyethylene glycol 600
Polyethylene glycol 600
Thermo Scientific Chemicals
Polyethylene glycol 600
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21798.30250 g
Catalog number ALFB21798.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene glycol 600
CAS25322-68-3
Health Hazard 1H227-H303-H315-H320-H333-H335
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
Melting Point17°C to 23°C
View more
Polyethylene glycol is used as a humectant in inks and adhesives. It acts as a solvent in soldering flux and as a dye carrier in inks. It is also useful as a modifier and binder in latex paint. It is used as an additive in synthetic lubricants, cutting oil and hydraulic fluids. It plays an important role for the precipitation of proteins.