Nonanoic acid, 97%
Nonanoic acid, 97%
Nonanoic acid, 97%
Thermo Scientific Chemicals
Nonanoic acid, 97%
CAS: 112-05-0 | C9H18O2 | 158.241 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21568.AK250 mL
Catalog number ALFB21568.AK
Price (MYR)
260.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
260.00
EA
Add to cart
Specifications
Chemical Name or MaterialNonanoic acid
CAS112-05-0
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Nonanoic acid is used in the preparation of plasticizers and lacquers. It is commonly used in conjunction with glyphosate, for a quick burn-down effect in the control of weeds in turfgrass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nonanoic acid is used in the preparation of plasticizers and lacquers. It is commonly used in conjunction with glyphosate, for a quick burn-down effect in the control of weeds in turfgrass.
Solubility
Soluble in water at 20°C,0.26g/L. Soluble in alcohol and dipropylene glycol.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Zhiyong Sun, et al. Carbon-limited fed-batch production of medium-chain-length polyhydroxyalkanoates from nonanoic acid by Pseudomonas putida KT2440.Appl. Microbiol. Biotechnol.,200774(1), 69-77.
  2. Madhu Aneja, et al. Trichoderma harzianum produces nonanoic acid, an inhibitor of spore germination and mycelial growth of two cacao pathogens.Physiological and Molecular Plant Pathology.,200567(6), 304-307.