1,3,5-Triphenylbenzene, 99+%
1,3,5-Triphenylbenzene, 99+%
1,3,5-Triphenylbenzene, 99+%
1,3,5-Triphenylbenzene, 99+%
Thermo Scientific Chemicals
1,3,5-Triphenylbenzene, 99+%
CAS: 612-71-5 | C24H18 | 306.41 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21353.1850 g
Catalog number ALFB21353.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3,5-Triphenylbenzene
CAS612-71-5
Melting Point172°C to 174°C
Recommended StorageAmbient temperatures
RTECS NumberWZ6530000
View more
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.
Solubility
Solubility in toluene is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Y. C. Lin and D. E. Williams. 1,3,5-Triphenylbenzene.Acta Cryst.1975B31318-320.
  2. Sakan Sirilaksanapong, et al. 1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.Chem. Commun., 201248(2), 293-295.