2-Ethyl-1-butanol, 99%
2-Ethyl-1-butanol, 99%
2-Ethyl-1-butanol, 99%
2-Ethyl-1-butanol, 99%
Thermo Scientific Chemicals
2-Ethyl-1-butanol, 99%
CAS: 97-95-0 | C6H14O | 102.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21026.AP500 mL
Catalog number ALFB21026.AP
Price (MYR)
2,346.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
2,346.00
EA
Add to cart
Specifications
Chemical Name or Material2-Ethyl-1-butanol
CAS97-95-0
Health Hazard 1H226-H302+H312
Health Hazard 2GHS H Statement
H311-H226-H302
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P312-P330-P363-P370+P378q-P501c
View more
It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.
Solubility
Soluble in water (4 mg/ml at 25°C), alcohol, ether, and most organic solvents.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. McGinty D. Fragrance material review on 2-ethyl-1-butanol.Food Chem Toxicol.201048(4), 85-88.
  2. Fumio Kimura and George C. Excess volumes of binary mixtures of 2-ethyl-1-butanol with hexane isomers at 298.15 K.Fluid Phase Equilib.198416(1), 77-86.