Furazolidone, 98%
Furazolidone, 98%
Furazolidone, 98%
Furazolidone, 98%
CAS: 67-45-8 | C8H7N3O5 | 225.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number ALFB20834.36
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFurazolidone
CAS67-45-8
Health Hazard 1H341
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
Monoamine oxidase inhibitor