1-Iodopentane, 98%, stab with copper
1-Iodopentane, 98%, stab with copper
1-Iodopentane, 98%, stab with copper
1-Iodopentane, 98%, stab with copper
Thermo Scientific Chemicals
1-Iodopentane, 98%, stab with copper
CAS: 628-17-1 | C5H11I | 198.047 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20781.22100 g
Catalog number ALFB20781.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Iodopentane
Name Notestabilized with copper
CAS628-17-1
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. F. Grane. Measurements of Transverse Dispersion in Granular Media. J. Chem. Eng. Data. 1961, 6 (2), 283-287 .
  2. Margaret Shea Chiles.; Donald D. Jackson.; Perry C. Reeves. Preparation and synthetic utility of phase-transfer catalysts anchored to polystyrene.J. Org. Chem. 1980, 45 (14),2915-2918 .