2,2-Dimethyl-3-octanol, 98%, Thermo Scientific™
2,2-Dimethyl-3-octanol, 98%, Thermo Scientific™
2,2-Dimethyl-3-octanol, 98%, Thermo Scientific™
2,2-Dimethyl-3-octanol, 98%, Thermo Scientific™
2,2-Dimethyl-3-octanol, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFB20410.1425 g
Catalog number ALFB20410.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2-Dimethyl-3-octanol
CAS19841-72-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.