3-Hexanol, 98%
3-Hexanol, 98%
3-Hexanol, 98%
Thermo Scientific Chemicals
3-Hexanol, 98%
CAS: 623-37-0 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB20210.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Hexanol
CAS623-37-0
Health Hazard 1H226-H302-H319
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
View more
It finds it application as a flavoring agent or adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds it application as a flavoring agent or adjuvant.
Solubility
Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Ziółkowska NE, et al. New active HIV-1 protease inhibitors derived from 3-hexanol: conformation study of the free inhibitors in crystalline state and in complex with the enzyme.Chem Biol Drug Des.2012,79(5), 798-809.
  2. Jörg Friedrich.; Gerhard M Schneider. Near-infrared spectroscopic investigations on phase behaviour and association of 1-octadecanol, 3-hexanol, and 3-methyl-3-pentanol in carbon dioxide and chlorotrifluoromethane.The Journal of Chemical Thermodynamics.1989,21(3), 307-319.