trans-Aconitic acid, 98%
trans-Aconitic acid, 98%
trans-Aconitic acid, 98%
trans-Aconitic acid, 98%
Thermo Scientific Chemicals
trans-Aconitic acid, 98%
CAS: 4023-65-8 | C6H6O6 | 174.108 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB20087.22
Price (MYR)
1,370.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,370.00
EA
Add to cart
Specifications
Chemical Name or Materialtrans-Aconitic acid
CAS4023-65-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It plays an important role as an antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes. trans-Aconitic acid is also used in synthesis of aconitate esters. It may be used for cross-linking polyvinyl alcohol film for preparing composite nanofiltration membranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It plays an important role as an antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes. trans-Aconitic acid is also used in synthesis of aconitate esters. It may be used for cross-linking polyvinyl alcohol film for preparing composite nanofiltration membranes.
Solubility
Soluble in water at 20°C 400g/L. Slightly soluble in ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Piang-Siong W, et al. Effect of catalytic conditions on the synthesis of new aconitate esters.Ind. Crops Prod.2012,35(1), 203-210.
  2. Dlamini DS, et al. Effect of Cross-Linking Agent Chemistry and Coating Conditions on Physical, Chemical and Separation Properties of PVA-PSf Composite Membranes.Sep. Sci. Technol.,2013.