2,5-Dimethyl-1,5-hexadien-3-ol, 97%
2,5-Dimethyl-1,5-hexadien-3-ol, 97%
2,5-Dimethyl-1,5-hexadien-3-ol, 97%
2,5-Dimethyl-1,5-hexadien-3-ol, 97%
Thermo Scientific Chemicals
2,5-Dimethyl-1,5-hexadien-3-ol, 97%
CAS: 17123-63-6 | C8H14O | 126.199 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20072.065 g
Catalog number ALFB20072.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2,5-Dimethyl-1,5-hexadien-3-ol
CAS17123-63-6
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC8H14O