Check Order Status
2-Propanol (USP), Fisher Chemical™
Element Search
Structure Search
Fisherbrand
2-Propanol (USP), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
ALFA516-200
200 L
Steel Drum
Catalog number
ALFA516-200
Price (MYR)
-
Quantity:
200 L
Packaging:
Steel Drum
Request bulk or custom format
Specifications
Chemical Name or Material
2-Propanol
pH
7
Cleanup Symbol
Solvent
Melting Point
-89.5°C
CAS
67-63-0
View more