Check Order Status
2-Propanol, ACS Grade, Fisher Chemical™
Element Search
Structure Search
2-Propanol, ACS Grade, Fisher Chemical™
In addition to meeting or exceeding the latest ACS specifications, are analyzed for more than 16 metals; Actual lot of analysis on label
Have Questions?
Catalog Number
Quantity
Packaging
ALFA416-4
4 L
Glass Bottle
Catalog number
ALFA416-4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
2-Propanol
pH
7
Cleanup Symbol
Solvent
Melting Point
-89.5°C
CAS
67-63-0
View more